Cyclopropanecarboxaldehyde ≥97.0% (by GC)

Supplier: TCI America

Synonyms: Cyclopropanecarbaldehyde

C1707-1G C1707-5G
TCC1707-5GEA 113.71 USD
TCC1707-5G TCC1707-1G
Cyclopropanecarboxaldehyde ≥97.0% (by GC)
Cyclopropanecarboxaldehyde

CAS Number: 1489-69-6
MDL Number: MFCD00012261
Molecular Formula: C4H6O
Molecular Weight: 70.09
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 102
Flash Point (°C): 7
Specific Gravity (20/20): 0.93
Storage Temperature: 0-10°C

Formula: C₄H₆O
MW: 70.09 g/mol
Boiling Pt: 102 °C
Density: 0.93 (20 °C)
Flash Pt: 7 °C
MDL Number: MFCD00012261
CAS Number: 1489-69-6
https://us.vwr.com/stibo/web/std.lang.all/89/11/28938911.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR