2-Chloro-1,3,2-benzodioxaborole (β-bromocatecholborane) ≥96.0% (by titrimetric analysis), Technical Grade
Supplier: TCI America
Synonyms:
2-Chloro-1,3,2-benzodioxaborole
, B-Chlorocatecholborane
C1669-5G
C1669-25G
TCC1669-25GEA
619.98
USD
TCC1669-25G
TCC1669-5G
2-Chloro-1,3,2-benzodioxaborole (β-bromocatecholborane) ≥96.0% (by titrimetric analysis), Technical Grade
2-Chloro-1,3,2-benzodioxaborole (β-Bromocatecholborane)
CAS Number: 55718-76-8
MDL Number: MFCD00043395
Molecular Formula: C6H4BClO2
Molecular Weight: 154.36
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 59
Flash Point (°C): 60
Other Product Names / Synonyms: 2-Chloro-1,3,2-benzodioxaborole
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...