Methyl (R)-(+)-2-chloropropionate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: (R)-(+)-2-Chloropropionic Acid Methyl Ester

C1633-5G
TCC1633-5GEA 110.64 USD
TCC1633-5G
Methyl (R)-(+)-2-chloropropionate ≥98.0% (by GC)
Methyl (R)-(+)-2-chloropropionate

CAS Number: 77287-29-7
MDL Number: MFCD00062940
Molecular Formula: C4H7ClO2
Molecular Weight: 122.55
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 134
Flash Point (°C): 37
Specific Gravity (20/20): 1.14
Specific rotation [a]20/D: 28 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Materials Chemistry, Precise Polymerization Reagents
Other Product Names / Synonyms: (R)-(+)-2-Chloropropionic Acid Methyl Ester

Formula: C₄H₇ClO₂
MW: 122.55 g/mol
Boiling Pt: 134 °C
Density: 1.14 (20 °C)
Flash Pt: 37 °C
MDL Number: MFCD00062940
CAS Number: 77287-29-7
https://us.vwr.com/stibo/web/std.lang.all/88/47/28938847.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR