2-Chloro-5-fluorotoluene ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 1-Chloro-4-fluoro-2-methylbenzene

C1537-25G C1537-5G
TCC1537-25GEA 40 USD
TCC1537-25G TCC1537-5G
2-Chloro-5-fluorotoluene ≥97.0% (by GC)
2-Chloro-5-fluorotoluene

CAS Number: 33406-96-1
MDL Number: MFCD00040853
Molecular Formula: C7H6ClF
Molecular Weight: 144.57
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.20

Formula: C₇H₆ClF
MW: 144.58 g/mol
Density: 1.20 (20 °C)
MDL Number: MFCD00040853
CAS Number: 33406-96-1
https://us.vwr.com/stibo/web/std.lang.all/87/55/28938755.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR