(S)-(+)-α-Cyclohexanemethylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: (S)-1-Cyclohexylethanamine(S)-(+)-1-Cyclohexylethylamine

C1531-5G C1531-25G
TCC1531-5GEA 61.37 USD
TCC1531-5G TCC1531-25G
(S)-(+)-α-Cyclohexanemethylamine ≥98.0% (by GC, titration analysis)
(S)-(+)-α-Cyclohexanemethylamine

CAS Number: 17430-98-7
MDL Number: MFCD00066261
Molecular Formula: C8H17N
Molecular Weight: 127.23
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 74
Flash Point (°C): 52
Specific Gravity (20/20): 0.87
Specific rotation [a]20/D: 3.3 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-(+)-(1-Aminoethyl)cyclohexane

Formula: C₈H₁₇N
MW: 127.2273 g/mol
Boiling Pt: 74 °C
Density: 0.87 (20 °C)
Flash Pt: 52 °C
MDL Number: MFCD00066261
CAS Number: 17430-98-7
https://us.vwr.com/stibo/web/std.lang.all/87/51/28938751.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR