Ethyl-6-(chloroformyl)hexanoate ≥97.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Ethyl 7-chloro-7-oxoheptanoate
C1414-5G
TCC1414-5GEA
231.94
USD
TCC1414-5G
Ethyl-6-(chloroformyl)hexanoate ≥97.0% (by titrimetric analysis)
Ethyl 6-(chloroformyl)hexanoate
CAS Number: 14794-32-2
MDL Number: MFCD00671507
Molecular Formula: C9H15ClO3
Molecular Weight: 206.67
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 141
Specific Gravity (20/20): 1.08
Other Product Names / Synonyms: 6-(Chloroformyl)hexanoic Acid Ethyl Ester, Ethyl 7-Chloro-7-oxoheptanoate
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...