Chloromethyl chloroformate ≥95.0% (by GC, titration analysis)
Supplier: TCI America
C1396-100G
C1396-25G
TCC1396-100GEA
522.57
USD
TCC1396-100G
TCC1396-25G
Chloromethyl chloroformate ≥95.0% (by GC, titration analysis)
Chloromethyl chloroformate
CAS Number: 22128-62-7
MDL Number: MFCD00077688
Molecular Formula: C2H2Cl2O2
Molecular Weight: 128.94
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 108
Flash Point (°C): 95
Specific Gravity (20/20): 1.45
Storage Temperature: 0-10°C
Other Product Names / Synonyms: Chloroformic Acid Chloromethyl Ester
Formula:
C₂H₂Cl₂O₂ Boiling Pt: 108 °C Density: 1.45 (20 °C) Flash Pt: 95 °C |
MDL Number:
MFCD00077688 CAS Number: 22128-62-7 UN: 2745 ADR: 6.1,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...