Cyclooctylamine ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Aminocyclooctane

C1223-25ML
TCC1223-25MLEA 142 USD
TCC1223-25ML
Cyclooctylamine ≥98.0% (by GC)
Cyclooctylamine

CAS Number: 5452-37-9
MDL Number: MFCD00001748
Molecular Formula: C8H17N
Molecular Weight: 127.23
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 190
Melting point (°C): -48
Flash Point (°C): 67
Specific Gravity (20/20): 0.91


Other Product Names / Synonyms: Aminocyclooctane

Formula: C₈H₁₇N
MW: 127.23 g/mol
Boiling Pt: 190 °C
Melting Pt: –48 °C
Density: 0.91 (20 °C)
Flash Pt: 67 °C
MDL Number: MFCD00001748
CAS Number: 5452-37-9
UN: 2735
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/84/68/28938468.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR