2-Chloro-6-fluorotoluene ≥98.0% (by GC)

Supplier: TCI America

C1181-25G
TCC1181-25GEA 30 USD
TCC1181-25G
2-Chloro-6-fluorotoluene ≥98.0% (by GC)
2-Chloro-6-fluorotoluene

CAS Number: 443-83-4
MDL Number: MFCD00000570
Molecular Formula: C7H6ClF
Molecular Weight: 144.57
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 153
Flash Point (°C): 46
Specific Gravity (20/20): 1.20

Formula: C₇H₆ClF
MW: 144.58 g/mol
Boiling Pt: 153 °C
Density: 1.20 (20 °C)
Flash Pt: 46 °C
MDL Number: MFCD00000570
CAS Number: 443-83-4
UN: 1993
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/84/33/28938433.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR