3,3-Diethoxypropiononitrile ≥95.0% (by GC)

Supplier: TCI America

Synonyms: 3,3-Diethoxypropanenitrile3,3-DiethoxypropionitrileCyanoacetaldehyde diethyl acetal

C1178-10ML
TCC1178-10MLEA 94.4 USD
TCC1178-10ML
3,3-Diethoxypropiononitrile ≥95.0% (by GC)
3,3-Diethoxypropiononitrile

CAS Number: 2032-34-0
MDL Number: MFCD00040558
Molecular Formula: C7H13NO2
Molecular Weight: 143.19
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 99
Flash Point (°C): 84
Specific Gravity (20/20): 0.95


Other Product Names / Synonyms: Cyanoacetaldehyde Diethyl Acetal

Formula: C₇H₁₃NO₂
MW: 143.19 g/mol
Boiling Pt: 99 °C
Density: 0.95 (20 °C)
Flash Pt: 84 °C
MDL Number: MFCD00040558
CAS Number: 2032-34-0
https://us.vwr.com/stibo/web/std.lang.all/84/30/28938430.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR