α,2-Dichloro-6-fluorotoluene ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
1-Chloro-2-(chloromethyl)-3-fluorobenzene
, 2-Chloro-6-fluorobenzyl chloride
C1081-25G
TCC1081-25GEA
86
USD
TCC1081-25G
α,2-Dichloro-6-fluorotoluene ≥98.0% (by GC)
α,2-Dichloro-6-fluorotoluene
CAS Number: 55117-15-2
MDL Number: MFCD00000896
Molecular Formula: C7H5Cl2F
Molecular Weight: 179.02
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 210
Flash Point (°C): 93
Specific Gravity (20/20): 1.37
Formula:
C₇H₅Cl₂F MW: 179.02 g/mol Boiling Pt: 210 °C Density: 1.37 (20 °C) Flash Pt: 93 °C |
MDL Number:
MFCD00000896 CAS Number: 55117-15-2 UN: 3265 ADR: 8,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...