2-Chlorobenzylamine ≥98.0% (by GC)

Supplier: TCI America

Synonyms: (2-Chlorophenyl)methanamine

C1065-25G
TCC1065-25GEA 35.53 USD
TCC1065-25G
2-Chlorobenzylamine ≥98.0% (by GC)
2-Chlorobenzylamine

CAS Number: 89-97-4
MDL Number: MFCD00008108
Molecular Formula: C7H8ClN
Molecular Weight: 141.60
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 219
Flash Point (°C): 88
Specific Gravity (20/20): 1.17

Formula: C₇H₈ClN
MW: 141.6 g/mol
Boiling Pt: 219 °C
Density: 1.17 (20 °C)
Flash Pt: 88 °C
MDL Number: MFCD00008108
CAS Number: 89-97-4
UN: 2735
ADR: 8,III
https://us.vwr.com/stibo/web/std.lang.all/83/28/28938328.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR