1-(4-Morpholino)cyclopentene ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 4-(1-Cyclopenten-1-yl)morpholine1-Morpholinocyclopentene4-(Cyclopent-1-en-1-yl)morpholine

C1040-25ML
TCC1040-25MLEA 188.73 USD
TCC1040-25ML
1-(4-Morpholino)cyclopentene ≥97.0% (by GC)
1-(4-Morpholino)cyclopentene

CAS Number: 936-52-7
MDL Number: MFCD00006162
Molecular Formula: C9H15NO
Molecular Weight: 153.23
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 254
Flash Point (°C): 60
Specific Gravity (20/20): 1.03


Other Product Names / Synonyms: N-(1-Cyclopenten-1-yl)morpholine

Formula: C₉H₁₅NO
MW: 153.22 g/mol
Boiling Pt: 254 °C
Density: 1.03 (20 °C)
Flash Pt: 60 °C
MDL Number: MFCD00006162
CAS Number: 936-52-7
https://us.vwr.com/stibo/web/std.lang.all/82/98/28938298.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR