Cyclobutyl methyl ketone ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
1-Cyclobutylethan-1-one
C1034-5ML
C1034-1ML
TCC1034-5MLEA
519
USD
TCC1034-5ML
TCC1034-1ML
Cyclobutyl methyl ketone ≥98.0% (by GC)
Cyclobutyl methyl ketone
CAS Number: 3019-25-8
MDL Number: MFCD00037148
Molecular Formula: C6H10O
Molecular Weight: 98.15
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 137
Flash Point (°C): 28
Specific Gravity (20/20): 0.90
Other Product Names / Synonyms: Cyclobutyl Methyl Ketone
Formula:
C₆H₁₀O MW: 98.15 g/mol Boiling Pt: 137 °C Density: 0.90 (20 °C) Flash Pt: 28 °C |
MDL Number:
MFCD00037148 CAS Number: 3019-25-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...