Chlorocyclobutane ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Cyclobutyl chloride

C1033-5ML C1033-1ML
TCC1033-5MLEA 315 USD
TCC1033-5ML TCC1033-1ML
Chlorocyclobutane ≥98.0% (by GC)
Chlorocyclobutane

CAS Number: 1120-57-6
MDL Number: MFCD00013736
Molecular Formula: C4H7Cl
Molecular Weight: 90.55
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 82
Flash Point (°C): -8
Specific Gravity (20/20): 0.99


Other Product Names / Synonyms: Cyclobutyl Chloride

Formula: C₄H₇Cl
MW: 90.55 g/mol
Boiling Pt: 82 °C
Density: 0.99 (20 °C)
Flash Pt: –8 °C
MDL Number: MFCD00013736
CAS Number: 1120-57-6
UN: 1127
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/82/90/28938290.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR