2-Chlorophenyl phosphorodichloridate ≥97.0% (by titrimetric analysis)

Supplier: TCI America

C0976-25G C0976-5G
TCC0976-25GEA 102.12 USD
TCC0976-25G TCC0976-5G
2-Chlorophenyl phosphorodichloridate ≥97.0% (by titrimetric analysis)
2-Chlorophenyl phosphorodichloridate

[Phosphorylating Agent]
CAS Number: 15074-54-1
MDL Number: MFCD00009704
Molecular Formula: C6H4Cl3O2P
Molecular Weight: 245.42
Purity/Analysis Method: 97.0% (T)
Form: Clear Liquid
Boiling point (°C): 256
Flash Point (°C): 105
Specific Gravity (20/20): 1.52


Product Categories: Synthetic Organic Chemistry, Phosphorylation, Nucleic Acid Synthesis
Other Product Names / Synonyms: 2-Chlorophenylphosphoryl Dichloride

Formula: C₆H₄Cl₃O₂P
Boiling Pt: 256 °C
Density: 1.52 (20 °C)
Flash Pt: 105 °C
MDL Number: MFCD00009704
CAS Number: 15074-54-1
UN: 3265
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/82/26/28938226.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR