Cyclopentanethiol ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Cyclopentyl mercaptan Mercaptocyclopentane

C0810-25ML C0810-5ML
TCC0810-5MLEA 43.54 USD
TCC0810-5ML TCC0810-25ML
Cyclopentanethiol ≥98.0% (by GC)
Cyclopentanethiol

CAS Number: 1679-07-8
MDL Number: MFCD00001369
Molecular Formula: C5H10S
Molecular Weight: 102.20
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 132
Flash Point (°C): 25
Specific Gravity (20/20): 0.96


Product Categories: Materials Chemistry, Self-Assembled Monolayer (SAM) Forming Agents
Other Product Names / Synonyms: Cyclopentyl Mercaptan, Mercaptocyclopentane

Formula: C₅H₁₀S
Boiling Pt: 132 °C
Density: 0.96 (20 °C)
Flash Pt: 25 °C
MDL Number: MFCD00001369
CAS Number: 1679-07-8
UN: 3336
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/78/47/28937847.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR