Cyanoacetohydrazide ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Cyanoacetic Acid HydrazideCyanoacetic HydrazideCyanoacetylhydrazine

C0800-25G
TCC0800-25GEA 79.17 USD
TCC0800-25G
Cyanoacetohydrazide ≥97.0% (by titrimetric analysis)
Cyanoacetohydrazide

CAS Number: 140-87-4
MDL Number: MFCD00007611
Molecular Formula: C3H5N3O
Molecular Weight: 99.09
Purity/Analysis Method: 97.0% (T)
Form: Crystal
Melting point (°C): 107


Other Product Names / Synonyms: Cyanoacetylhydrazine, Cyanoacetic Hydrazide, Cyanoacetic Acid Hydrazide

Formula: C₃H₅N₃O
MW: 99.09 g/mol
Melting Pt: 107 °C
MDL Number: MFCD00007611
CAS Number: 140-87-4
https://us.vwr.com/stibo/web/std.lang.all/78/36/28937836.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR