Cycloheptylamine ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Aminocycloheptane

C0679-10G
TCC0679-10GEA 109 USD
TCC0679-10G
Cycloheptylamine ≥97.0% (by GC, titration analysis)
Cycloheptylamine

CAS Number: 5452-35-7
MDL Number: MFCD00004153
Molecular Formula: C7H15N
Molecular Weight: 113.20
Purity/Analysis Method: 97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 170
Flash Point (°C): 47
Specific Gravity (20/20): 0.89


Other Product Names / Synonyms: Aminocycloheptane

Formula: C₇H₁₅N
MW: 113.2 g/mol
Boiling Pt: 170 °C
Density: 0.89 (20 °C)
Flash Pt: 47 °C
MDL Number: MFCD00004153
CAS Number: 5452-35-7
UN: 1993
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/77/18/28937718.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR