1-Chloro-2-fluorobenzene ≥98.0% (by GC)

Supplier: TCI America

C0647-25G C0647-10G
TCC0647-10GEA 36.54 USD
TCC0647-10G TCC0647-25G
1-Chloro-2-fluorobenzene ≥98.0% (by GC)
1-Chloro-2-fluorobenzene

CAS Number: 348-51-6
MDL Number: MFCD00000533
Molecular Formula: C6H4ClF
Molecular Weight: 130.55
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 138
Flash Point (°C): 31
Specific Gravity (20/20): 1.24


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Chlorofluorobenzene

Formula: C₆H₄ClF
MW: 130.55 g/mol
Boiling Pt: 138 °C
Density: 1.24 (20 °C)
Flash Pt: 31 °C
MDL Number: MFCD00000533
CAS Number: 348-51-6
https://us.vwr.com/stibo/web/std.lang.all/76/87/28937687.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR