Cycloheptane ≥98.0% (by GC)

Supplier: TCI America

C0465-25ML C0465-5ML
TCC0465-5MLEA 43 USD
TCC0465-5ML TCC0465-25ML
Cycloheptane ≥98.0% (by GC)
Cycloheptane

CAS Number: 291-64-5
MDL Number: MFCD00004147
Molecular Formula: C7H14
Molecular Weight: 98.19
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 118
Flash Point (°C): 15
Specific Gravity (20/20): 0.81

Formula: C₇H₁₄
Boiling Pt: 118 °C
Density: 0.81 (20 °C)
Flash Pt: 15 °C
MDL Number: MFCD00004147
CAS Number: 291-64-5
UN: 1206
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/74/86/28937486.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR