Cyclopropanecarboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

C0387-100ML C0387-25ML C0387-500ML
TCC0387-100MLEA 68.01 USD
TCC0387-100ML TCC0387-25ML TCC0387-500ML
Cyclopropanecarboxylic acid ≥98.0% (by GC, titration analysis)
Cyclopropanecarboxylic acid

CAS Number: 1759-53-1
MDL Number: MFCD00001287
Molecular Formula: C4H6O2
Molecular Weight: 86.09
Purity/Analysis Method: 98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 183
Flash Point (°C): 71
Freezing point (°C): 16
Specific Gravity (20/20): 1.09

Formula: C₄H₆O₂
MW: 86.09 g/mol
Boiling Pt: 183 °C
Density: 1.09 (20 °C)
Flash Pt: 71 °C
MDL Number: MFCD00001287
CAS Number: 1759-53-1
UN: 3265
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/74/12/28937412.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR