4-Chloro-3-nitrotoluene ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 1-Chloro-4-methyl-2-nitrobenzene

C0232-25G
TCC0232-25GEA 45 USD
TCC0232-25G
4-Chloro-3-nitrotoluene ≥97.0% (by GC)
4-Chloro-3-nitrotoluene

CAS Number: 89-60-1
MDL Number: MFCD00007085
Molecular Formula: C7H6ClNO2
Molecular Weight: 171.58
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 260
Flash Point (°C): 129
Specific Gravity (20/20): 1.31

Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Boiling Pt: 260 °C
Density: 1.31 (20 °C)
Flash Pt: 129 °C
MDL Number: MFCD00007085
CAS Number: 89-60-1
UN: 2433
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/72/86/28937286.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR