6,6'-Bis(chloromethyl)-2,2'-bipyridyl ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 6,6'-Bis(chloromethyl)-2,2'-bipyridine

B4645-200MG B4645-1G
TCB4645-200MGEA 135 USD
TCB4645-200MG TCB4645-1G
6,6'-Bis(chloromethyl)-2,2'-bipyridyl ≥98.0% (by GC, titration analysis)
6,6'-Bis(chloromethyl)-2,2'-bipyridyl

CAS Number: 74065-64-8
MDL Number: MFCD02181082
Molecular Formula: C12H10Cl2N2
Molecular Weight: 253.13
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 162


Product Categories: Synthetic Organic Chemistry, Ligands
Other Product Names / Synonyms: 6,6'-Bis(chloromethyl)-2,2'-bipyridine

Formula: C₁₂H₁₀Cl₂N₂
MW: 253.13 g/mol
Melting Pt: 162 °C
MDL Number: MFCD02181082
CAS Number: 74065-64-8
https://us.vwr.com/stibo/web/std.lang.all/59/12/28935912.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR