1,4-Benzenediboronic acid bis(pinacol)ester ≥98.0%
Supplier: TCI America
Synonyms:
1,4-Bis(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzene
CAS Number: 99770-93-1
MDL Number: MFCD08276852
Molecular Formula: C18H28B2O4
Molecular Weight: 330.04
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 243
Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives, Materials Chemistry, Building Blocks for Small Molecule Semiconductors, Conducting Polymer Research Reagents
Other Product Names / Synonyms: 1,4-Bis(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, 1,4-Phenylenebis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane)
Formula:
C₁₈H₂₈B₂O₄ MW: 330.04 g/mol Melting Pt: 243 °C |
MDL Number:
MFCD08276852 CAS Number: 99770-93-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...