Boc-Glu-OtBu ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Boc-Glu-OtBuN-(tert-Butoxycarbonyl)-L-glutamic Acid 1-tert-Butyl Ester1-tert-Butyl N-Boc-L-glutamateN-Boc-L-glutamic Acid 1-tert-Butyl Ester

B4534-5G B4534-25G
TCB4534-5GEA 51.77 USD
TCB4534-5G TCB4534-25G
Boc-Glu-OtBu ≥97.0% (by HPLC, titration analysis)
Boc-Glu-OtBu

CAS Number: 24277-39-2
MDL Number: MFCD00038273
Molecular Formula: C14H25NO6
Molecular Weight: 303.36
Purity/Analysis Method: 97.0% (HPLC,T)
Form: Crystal
Melting point (°C): 113
Specific rotation [a]20/D: -33 deg (C=0.6, MeOH)
Storage Temperature: 0°C


Product Categories: Bioscience, Amino Acids (N-Protected), Amino Acids (C-Protected)
Other Product Names / Synonyms: N-(tert-Butoxycarbonyl)-L-glutamic Acid 1-tert-Butyl Ester, 1-tert-Butyl N-Boc-L-glutamate, N-Boc-L-glutamic Acid 1-tert-Butyl Ester, Boc-Glu-OtBu

Formula: C14H25NO6
MW: 303.36 g/mol
Melting Pt: 113 °C
MDL Number: MFCD00038273
CAS Number: 24277-39-2
https://us.vwr.com/stibo/web/std.lang.all/58/12/28935812.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR