1,3-Bis(4-chlorophenyl)thiourea ≥98.0% (by HPLC)
Supplier: TCI America
B4481-1G
TCB4481-1GEA
343
USD
TCB4481-1G
1,3-Bis(4-chlorophenyl)thiourea ≥98.0% (by HPLC)
1,3-Bis(4-chlorophenyl)thiourea
CAS Number: 1220-00-4
MDL Number: MFCD00275761
Molecular Formula: C13H10Cl2N2S
Molecular Weight: 297.20
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 172
Formula:
C₁₃H₁₀Cl₂N₂S Melting Pt: 172 °C |
MDL Number:
MFCD00275761 CAS Number: 1220-00-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...