Betaxolol hydrochloride ≥98.0% (by HPLC)

Supplier: TCI America

B4474-50MG B4474-250MG
TCB4474-250MGEA 422.12 USD
TCB4474-250MG TCB4474-50MG
Betaxolol hydrochloride ≥98.0% (by HPLC)
Betaxolol hydrochloride

CAS Number: 63659-19-8
MDL Number: MFCD00242959
Molecular Formula: C18H29NO3
Molecular Weight: 343.89
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 115

Formula: C₁₈H₂₉NO₃
Melting Pt: 115 °C
MDL Number: MFCD00242959
CAS Number: 63659-19-8
https://us.vwr.com/stibo/web/std.lang.all/57/59/28935759.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR