4-Bromo-1-naphthaleneboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: (4-Bromonaphthalen-1-yl)boronic acid

B4463-1G
TCB4463-1GEA 66 USD
TCB4463-1G
4-Bromo-1-naphthaleneboronic acid (contains varying amounts of Anhydride)
4-Bromo-1-naphthaleneboronic acid

CAS Number: 145965-14-6
MDL Number: MFCD08701764
Molecular Formula: C10H8BBrO2
Molecular Weight: 250.89
Form: Crystal
Color: White

Formula: C₁₀H₈BBrO₂
MW: 250.88 g/mol
MDL Number: MFCD08701764
CAS Number: 145965-14-6
https://us.vwr.com/stibo/web/std.lang.all/57/48/28935748.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR