8-Bromo-7-(2-butyn-1-yl)-3-methylxanthine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
8-Bromo-7-(2-butyn-1-yl)-3,7-dihydro-3-methyl-1H-purine-2,6-dione
CAS Number: 666816-98-4
MDL Number: MFCD22126091
Molecular Formula: C10H9BrN4O2
Molecular Weight: 297.11
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 285
Product Categories: Bioscience, Nucleobases and Their Analogs
Other Product Names / Synonyms: 8-Bromo-7-(2-butyn-1-yl)-3,7-dihydro-3-methyl-1H-purine-2,6-dione
Formula:
C₁₀H₉BrN₄O₂ MW: 297.11 g/mol Melting Pt: 285 °C |
MDL Number:
MFCD22126091 CAS Number: 666816-98-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...