2-Bromo-9,9-bis(2-ethylhexyl)fluorene ≥98.0% (by GC)

Supplier: TCI America

B4441-1G
TCB4441-1GEA 277.02 USD
TCB4441-1G
2-Bromo-9,9-bis(2-ethylhexyl)fluorene ≥98.0% (by GC)
2-Bromo-9,9-bis(2-ethylhexyl)fluorene

CAS Number: 355135-07-8
MDL Number: MFCD12024275
Molecular Formula: C29H41Br
Molecular Weight: 469.55
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.09

Formula: C₂₉H₄₁Br
MW: 469.55 g/mol
Density: 1.09 (20 °C)
MDL Number: MFCD12024275
CAS Number: 355135-07-8
https://us.vwr.com/stibo/web/std.lang.all/57/29/28935729.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR