5-Bromo-4-chloro-2-(methylthio)pyrimidine ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 5-bromo-4-chloro-2-(methylsulfanyl)pyrimidine5-bromo-4-chloro-2-(methylsulphanyl)pyrimidine

B4406-1G
TCB4406-1GEA 60 USD
TCB4406-1G
5-Bromo-4-chloro-2-(methylthio)pyrimidine ≥98.0% (by GC)
5-Bromo-4-chloro-2-(methylthio)pyrimidine

CAS Number: 63810-78-6
MDL Number: MFCD00023232
Molecular Formula: C5H4BrClN2S
Molecular Weight: 239.52
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 47

Formula: C₅H₄BrClN₂S
MW: 239.5200 g/mol
Melting Pt: 47 °C
MDL Number: MFCD00023232
CAS Number: 63810-78-6
https://us.vwr.com/stibo/web/std.lang.all/57/00/28935700.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR