(tert-Butyldimethylsilyl)acetylene ≥97.0% (by GC)

Supplier: TCI America

B4365-5G B4365-25G
TCB4365-25GEA 576.52 USD
TCB4365-25G TCB4365-5G
(tert-Butyldimethylsilyl)acetylene ≥97.0% (by GC)
(tert-Butyldimethylsilyl)acetylene

CAS Number: 86318-61-8
MDL Number: MFCD00191877
Molecular Formula: C8H16Si
Molecular Weight: 140.30
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 110
Flash Point (°C): 6
Specific Gravity (20/20): 0.77


Product Categories: Synthetic Organic Chemistry, Click Chemistry, Building Blocks ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: (tert-Butyl)ethynyldimethylsilane

Formula: C₈H₁₆Si
MW: 140.3 g/mol
Boiling Pt: 110 °C
Density: 0.77 (20 °C)
Flash Pt: 6 °C
MDL Number: MFCD00191877
CAS Number: 86318-61-8
https://us.vwr.com/stibo/web/std.lang.all/56/57/28935657.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR