Bis(4-dimethylaminodithiobenzil)nickel(II) ≥93.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
Bis[1-(4-dimethylamino)phenyl-2-phenyl-1,2-ethenedithiolato]nickel(II)
B4361-1G
TCB4361-1GEA
261.75
USD
TCB4361-1G
Bis(4-dimethylaminodithiobenzil)nickel(II) ≥93.0% (by HPLC, titration analysis)
Bis(4-dimethylaminodithiobenzil)nickel(II)
CAS Number: 38465-55-3
MDL Number: MFCD00135960
Molecular Formula: C32H30N2NiS4
Molecular Weight: 629.54
Purity/Analysis Method:
Form: Crystal
Product Categories: Materials Chemistry, Organic Transistor Materials, Magnetic Metal Complexes (Magnetic Materials), Near Infrared Absorption Materials
Other Product Names / Synonyms: Bis[1-(4-dimethylamino)phenyl-2-phenyl-1,2-ethenedithiolato]nickel(II), BDN
Formula:
C₃₂H₃₀N₂NiS₄ MW: 629.54 g/mol |
MDL Number:
MFCD00135960 CAS Number: 38465-55-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...