Butyl-O-butyryllactate ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
2-(Butyryloxy)propionic Acid Butyl Ester
, O-Butyryllactic Acid Butyl Ester
, Butyl 2-(Butyryloxy)propionate
, Butyl Butyryllactate
CAS Number: 7492-70-8
MDL Number: MFCD00027213
Molecular Formula: C11H20O4
Molecular Weight: 216.28
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 90
Flash Point (°C): 113
Specific Gravity (20/20): 0.98
Other Product Names / Synonyms: 2-(Butyryloxy)propionic Acid Butyl Ester, Butyl 2-(Butyryloxy)propionate, O-Butyryllactic Acid Butyl Ester
Formula:
C₁₁H₂₀O₄ MW: 216.28 g/mol Boiling Pt: 90 °C Density: 0.98 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00027213 CAS Number: 7492-70-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...