5'-Bromo-m-terphenyl ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-Bromo-3,5-diphenylbenzene5'-Bromo-1,1':3',1''-terphenyl

B4308-5G B4308-25G
TCB4308-25GEA 247.58 USD
TCB4308-25G TCB4308-5G
5'-Bromo-m-terphenyl ≥98.0% (by GC)
5'-Bromo-m-terphenyl

CAS Number: 103068-20-8
MDL Number: MFCD00196170
Molecular Formula: C18H13Br
Molecular Weight: 309.21
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 111
Lambda max.: 252 nm (CH2Cl2)


Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: 1-Bromo-3,5-diphenylbenzene, 5'-Bromo-1,1':3',1''-terphenyl

Formula: C₁₈H₁₃Br
MW: 309.21 g/mol
Melting Pt: 111 °C
MDL Number: MFCD00196170
CAS Number: 103068-20-8
https://us.vwr.com/stibo/web/std.lang.all/56/09/28935609.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR