5-Bromo-2-iodobenzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
B4296-5G
TCB4296-5GEA
47.71
USD
TCB4296-5G
5-Bromo-2-iodobenzoic acid ≥98.0% (by GC, titration analysis)
5-Bromo-2-iodobenzoic acid
CAS Number: 21740-00-1
MDL Number: MFCD00079717
Molecular Formula: C7H4BrIO2
Molecular Weight: 326.92
Purity/Analysis Method:
Melting point (°C): 162
Formula:
C₇H₄BrIO₂ Melting Pt: 162 °C |
MDL Number:
MFCD00079717 CAS Number: 21740-00-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...