1,3,5-Benzenetricarbonitrile ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1,3,5-Tricyanobenzene

B4294-5G B4294-1G
TCB4294-5GEA 473.87 USD
TCB4294-5G TCB4294-1G
1,3,5-Benzenetricarbonitrile ≥98.0% (by GC)
1,3,5-Benzenetricarbonitrile

CAS Number: 10365-94-3
MDL Number: MFCD00019746
Molecular Formula: C9H3N3
Molecular Weight: 153.14
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 259


Other Product Names / Synonyms: 1,3,5-Tricyanobenzene

Formula: C₉H₃N₃
MW: 153.14 g/mol
Melting Pt: 259 °C
MDL Number: MFCD00019746
CAS Number: 10365-94-3
https://us.vwr.com/stibo/web/std.lang.all/55/98/28935598.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR