N-Boc-N-methylethylenediamine ≥95.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
N-Boc-(2-Aminoethyl)-N-methylamine
, N-Methylethane-1,2-diamine, N-BOC protected
B4277-1ML
B4277-5ML
TCB4277-1MLEA
44.66
USD
TCB4277-1ML
TCB4277-5ML
N-Boc-N-methylethylenediamine ≥95.0% (by GC, titration analysis)
N-Boc-N-methyl-ethylenediamine
CAS Number: 121492-06-6
MDL Number: MFCD01317789
Molecular Formula: C8H18N2O2
Molecular Weight: 174.24
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 79
Flash Point (°C): 100
Specific Gravity (20/20): 0.98
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...