Brinzolamide ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: (R)-4-(Ethylamino)-2-(3-methoxypropyl)-3,4-dihydro-2H-thieno[3,2-e][1,2]thiazine-6-sulfonamide 1,1-Dioxide

B4258-25MG B4258-100MG
TCB4258-25MGEA 70.04 USD
TCB4258-25MG TCB4258-100MG
Brinzolamide ≥98.0% (by HPLC)
Brinzolamide

CAS Number: 138890-62-7
MDL Number: MFCD08067749
Molecular Formula: C12H21N3O5S3
Molecular Weight: 383.50
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 132
Storage Temperature: 0°C


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Other Pharmaceutical Ingredients
Other Product Names / Synonyms: (R)-4-(Ethylamino)-2-(3-methoxypropyl)-3,4-dihydro-2H-thieno[3,2-e][1,2]thiazine-6-sulfonamide 1,1-Dioxide

Formula: C₁₂H₂₁N₃O₅S₃
MW: 383.50 g/mol
Melting Pt: 132 °C
MDL Number: MFCD08067749
CAS Number: 138890-62-7
https://us.vwr.com/stibo/web/std.lang.all/55/74/28935574.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR