Coumarin 545T ≥98.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: 10-(2-Benzothiazolyl)-2,3,6,7-tetrahydro-1,1,7,7-tetramethyl-1H,5H,11H-[1]benzopyrano[6,7,8-ij]quinolizin-11-one

B4257-200MG
TCB4257-200MGEA 327.75 USD
TCB4257-200MG
Coumarin 545T ≥98.0% (by total nitrogen basis)
Coumarin 545T

CAS Number: 155306-71-1
MDL Number: MFCD03427187
Molecular Formula: C26H26N2O2S
Molecular Weight: 430.57
Purity/Analysis Method: 98.0% (N)
Form: Crystal
Melting point (°C): 228


Product Categories: Materials Chemistry, Organic Light-Emitting Diode (OLED) Materials, Coumarin Dyes (Functional Organic Dyes)
Other Product Names / Synonyms: 10-(2-Benzothiazolyl)-2,3,6,7-tetrahydro-1,1,7,7-tetramethyl-1H,5H,11H-[1]benzopyrano[6,7,8-ij]quinolizin-11-one

Formula: C₂₆H₂₆N₂O₂S
MW: 430.57 g/mol
Melting Pt: 228 °C
MDL Number: MFCD03427187
CAS Number: 155306-71-1
https://us.vwr.com/stibo/web/std.lang.all/55/72/28935572.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR