Bis(4-chloro-2-nitrophenyl)disulfide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 4,4'-Dichloro-2,2'-dinitrodiphenyl Disulfide4,4'-Dichloro-2,2'-dinitrodiphenyl Disulphide

B4247-25G B4247-5G
TCB4247-5GEA 72.47 USD
TCB4247-5G TCB4247-25G
Bis(4-chloro-2-nitrophenyl)disulfide ≥98.0% (by HPLC, total nitrogen)
Bis(4-chloro-2-nitrophenyl)disulfide

CAS Number: 2050-66-0
MDL Number: MFCD00047692
Molecular Formula: C12H6Cl2N2O4S2
Molecular Weight: 377.21
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 216


Other Product Names / Synonyms: 4,4'-Dichloro-2,2'-dinitrodiphenyl Disulfide

Formula: C₁₂H₆Cl₂N₂O₄S₂
MW: 377.21 g/mol
Melting Pt: 216 °C
MDL Number: MFCD00047692
CAS Number: 2050-66-0
https://us.vwr.com/stibo/web/std.lang.all/55/64/28935564.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR