2,6-Bis[(2S,5S)-4,4-diphenyl-1-aza-3-oxabicyclo[3.3.0]octan-2-yl]pyridine ≥90.0% (by HPLC)
Supplier: TCI America
B4228-50MG
TCB4228-50MGEA
168
USD
TCB4228-50MG
2,6-Bis[(2S,5S)-4,4-diphenyl-1-aza-3-oxabicyclo[3.3.0]octan-2-yl]pyridine ≥90.0% (by HPLC)
2,6-Bis[(2S,5S)-4,4-diphenyl-1-aza-3-oxabicyclo[3.3.0]octan-2-yl]pyridine
CAS Number: 1450841-27-6
Molecular Formula: C41H39N3O2
Molecular Weight: 605.78
Purity/Analysis Method:
Form: Crystal
Specific rotation [a]20/D: -174 deg (C=1, CHCl3)
Formula:
C₄₁H₃₉N₃O₂ |
CAS Number:
1450841-27-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...