Butyraldehyde dibutyl acetal ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
Butyraldehyde Dibutyl Acetal
, 1,1-Dibutoxybutane
B4218-5G
B4218-25G
TCB4218-5GEA
103.5
USD
TCB4218-5G
TCB4218-25G
Butyraldehyde dibutyl acetal ≥98.0% (by GC)
Butyraldehyde dibutyl acetal
CAS Number: 5921-80-2
MDL Number: MFCD00561028
Molecular Formula: C12H26O2
Molecular Weight: 202.34
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 214
Specific Gravity (20/20): 0.84
Other Product Names / Synonyms: 1,1-Dibutoxybutane
Formula:
C₁₂H₂₆O₂ MW: 202.34 g/mol Boiling Pt: 214 °C Density: 0.84 (20 °C) |
MDL Number:
MFCD00561028 CAS Number: 5921-80-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...