4-Bromobenzamide ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
B4216-25G
B4216-5G
TCB4216-25GEA
145.15
USD
TCB4216-25G
TCB4216-5G
4-Bromobenzamide ≥98.0% (by HPLC, total nitrogen)
4-Bromobenzamide
CAS Number: 698-67-9
MDL Number: MFCD00007991
Molecular Formula: C7H6BrNO
Molecular Weight: 200.04
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 193
Lambda max.: 242 nm (H2SO4 aq.)
Formula:
C₇H₆BrNO MW: 200.03 g/mol Melting Pt: 193 °C |
MDL Number:
MFCD00007991 CAS Number: 698-67-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...