Bis(acetato)aqua[(S,S)-4,6-bis(4-isopropyl-2-oxazolin-2-yl)-m-xylene]rhodium
Supplier: TCI America
B4195-10MG
TCB4195-10MGEA
328.72
USD
TCB4195-10MG
Bis(acetato)aqua[(S,S)-4,6-bis(4-isopropyl-2-oxazolin-2-yl)-m-xylene]rhodium
Bis(acetato)aqua[(S,S)-4,6-bis(4-isopropyl-2-oxazolin-2-yl)-m-xylene]rhodium
CAS Number: 929896-28-6
Molecular Formula: C24H35N2O7Rh
Molecular Weight: 566.46
Form: Crystal
Color: Slightly Pale Yellow
Formula:
C₂₄H₃₅N₂O₇Rh |
CAS Number:
929896-28-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...