(R)-1,1'-Binaphthyl-2,2'-disulfonyl dichloride ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: (R)-1,1'-Binaphthyl-2,2'-disulfonyl Chloride(R)-1,1'-Binaphthyl-2,2'-disulphonyl Chloride

B4187-200MG B4187-1G
TCB4187-1GEA 931.5 USD
TCB4187-1G TCB4187-200MG
(R)-1,1'-Binaphthyl-2,2'-disulfonyl dichloride ≥98.0% (by HPLC)
(R)-1,1'-Binaphthyl-2,2'-disulfonyl dichloride

CAS Number: 1187629-43-1
Molecular Formula: C20H12Cl2O4S2
Molecular Weight: 451.33
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 235
Specific rotation [a]20/D: -44 deg (C=1, THF)


Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis
Other Product Names / Synonyms: (R)-1,1'-Binaphthyl-2,2'-disulfonyl Chloride

Formula: C₂₀H₁₂Cl₂O₄S₂
MW: 451.33 g/mol
Melting Pt: 235 °C
CAS Number: 1187629-43-1

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR