1-Bromo-3-hexylbenzene ≥98.0% (by GC)

Supplier: TCI America

B4180-5G
TCB4180-5GEA 90 USD
TCB4180-5G
1-Bromo-3-hexylbenzene ≥98.0% (by GC)
1-Bromo-3-hexylbenzene

CAS Number: 38409-59-5
MDL Number: MFCD09909567
Molecular Formula: C12H17Br
Molecular Weight: 241.17
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 98
Flash Point (°C): 120
Specific Gravity (20/20): 1.18


Other Product Names / Synonyms: 3-Hexylbromobenzene

Formula: C₁₂H₁₇Br
Boiling Pt: 98 °C
Density: 1.18 (20 °C)
Flash Pt: 120 °C
MDL Number: MFCD09909567
CAS Number: 38409-59-5
https://us.vwr.com/stibo/web/std.lang.all/55/17/28935517.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR