Bromopentamethylbenzene ≥97.0% (by GC)
Supplier: TCI America
Synonyms:
Pentamethylbromobenzene
, 1-Bromo-2,3,4,5,6-pentamethylbenzene
B4154-25G
B4154-5G
TCB4154-5GEA
57
USD
TCB4154-5G
TCB4154-25G
Bromopentamethylbenzene ≥97.0% (by GC)
Bromopentamethylbenzene
CAS Number: 5153-40-2
MDL Number: MFCD00013523
Molecular Formula: C11H15Br
Molecular Weight: 227.15
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 290
Melting point (°C): 162
Other Product Names / Synonyms: Pentamethylbromobenzene
Formula:
C₁₁H₁₅Br MW: 227.14 g/mol Boiling Pt: 290 °C Melting Pt: 162 °C |
MDL Number:
MFCD00013523 CAS Number: 5153-40-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...