Brimonidine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 5-Bromo-N-(2-imidazolin-2-yl)-6-quinoxalinamineUK 14,304

B4132-200MG B4132-1G
TCB4132-1GEA 492 USD
TCB4132-1G TCB4132-200MG
Brimonidine ≥98.0% (by HPLC, titration analysis)
Brimonidine

CAS Number: 59803-98-4
MDL Number: MFCD00153878
Molecular Formula: C11H10BrN5
Molecular Weight: 292.14
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 247


Product Categories: Bioscience, Other Pharmaceutical Ingredients
Other Product Names / Synonyms: 5-Bromo-N-(2-imidazolin-2-yl)-6-quinoxalinamine, UK 14,304

Formula: C₁₁H₁₀BrN₅
MW: 292.14 g/mol
Melting Pt: 247 °C
MDL Number: MFCD00153878
CAS Number: 59803-98-4
https://us.vwr.com/stibo/web/std.lang.all/54/59/28935459.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR